Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptlc1
Entrez Gene ID
361213
Gene Name
serine palmitoyltransferase, long chain base subunit 1
Gene Aliases
Lcb1, RGD1306617, Spt1
Chromosome Location
Chr.17: 11856525 - 11895566 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108406.1 NP_001101876.1 7 - 8 701 75

View Full Product Details