The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079941.1 | NP_001073410.1 | 4 - 5 | 1015 | 79 |
XM_006233835.2 | XP_006233897.1 | 4 - 5 | 1024 | 79 | |
GenBank mRNA | BC129109.1 | - | 4 - 5 | 1015 | 79 |