Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmpd1
Entrez Gene ID
313244
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 60752372 - 60837773 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238143.2 XP_006238205.1 4 - 5 1861 83
XM_006238144.2 XP_006238206.1 4 - 5 1814 83
XM_008763690.1 XP_008761912.1 4 - 5 569 83

View Full Product Details