Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf38
Entrez Gene ID
171501
Gene Name
ring finger protein 38
Gene Aliases
Oip1
Chromosome Location
Chr.5: 59606754 - 59713108 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134467.1 NP_604462.1 5 - 6 715 101
XM_006238012.2 XP_006238074.1 5 - 6 965 101
XM_006238014.2 XP_006238076.1 5 - 6 863 101
XM_006238015.2 XP_006238077.1 4 - 5 815 101
XM_006238016.2 XP_006238078.1 4 - 5 1062 101
XM_006238017.2 XP_006238079.1 4 - 5 1835 101
XM_006238018.2 XP_006238080.1 5 - 6 903 101
XM_006238020.2 XP_006238082.1 5 - 6 708 101
XM_008763664.1 XP_008761886.1 4 - 5 1489 101
XM_008763665.1 XP_008761887.1 4 - 5 1402 101
XM_008763666.1 XP_008761888.1 4 - 5 1387 101
XM_008763667.1 XP_008761889.1 6 - 7 858 101
XM_008763668.1 XP_008761890.1 5 - 6 801 101
GenBank mRNA AF480444.1 - 5 - 6 715 101
BC083548.1 - 4 - 5 630 101

View Full Product Details