Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc3
Entrez Gene ID
313243
Gene Name
exosome component 3
Gene Aliases
-
Chromosome Location
Chr.5: 60866498 - 60871737 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107936.1 NP_001101406.1 1 - 2 348 70
XM_006238067.2 XP_006238129.1 1 - 2 392 70

View Full Product Details