Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ripk2
Entrez Gene ID
362491
Gene Name
receptor-interacting serine-threonine kinase 2
Gene Aliases
-
Chromosome Location
Chr.5: 29839477 - 29870390 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191865.1 NP_001178794.1 3 - 4 623 119

View Full Product Details