Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glipr2
Entrez Gene ID
679819
Gene Name
GLI pathogenesis-related 2
Gene Aliases
-
Chromosome Location
Chr.5: 59415407 - 59446732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001054584.5 XP_001054584.2 1 - 2 113 72

View Full Product Details