The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133575.1 | NP_598259.1 | 10 - 11 | 1396 | 169 |
GenBank mRNA | U49066.1 | - | 10 - 11 | 1396 | 169 |