Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rftn2
Entrez Gene ID
363231
Gene Name
raftlin family member 2
Gene Aliases
RGD1307304
Chromosome Location
Chr.9: 61754859 - 61810964 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_343571.7 XP_343572.4 1 - 2 688 95

View Full Product Details