The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181383.2 | NP_852048.1 | 2 - 3 | 196 | 118 |
GenBank mRNA | AY280501.1 | - | 2 - 3 | 196 | 118 |