Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il1r2
Entrez Gene ID
117022
Gene Name
interleukin 1 receptor, type II
Gene Aliases
-
Chromosome Location
Chr.9: 46840642 - 46881239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053953.1 NP_446405.1 3 - 4 494 102
XM_008766983.1 XP_008765205.1 3 - 4 879 102
XM_008766984.1 XP_008765206.1 3 - 4 650 102
GenBank mRNA BC091564.1 - 3 - 4 468 102
FQ228742.1 - 3 - 4 493 102
FQ229102.1 - 3 - 4 493 102
Z22812.1 - 3 - 4 494 102

View Full Product Details