Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgol2
Entrez Gene ID
316425
Gene Name
shugoshin-like 2 (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.9: 64898420 - 64923023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271191.1 NP_001258120.1 6 - 7 3325 98
XM_006244988.2 XP_006245050.1 7 - 8 3602 98

View Full Product Details