Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nif3l1
Entrez Gene ID
301431
Gene Name
NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.9: 65329142 - 65349280 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244958.2 XP_006245020.1 6 - 7 1109 68
XM_006244960.2 XP_006245022.1 6 - 7 1080 68
XM_006244961.2 XP_006245023.1 6 - 7 1105 68
XM_006244962.2 XP_006245024.1 5 - 6 978 68
GenBank mRNA BC097992.1 - 6 - 7 1041 68

View Full Product Details