The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106567.1 | NP_001100037.1 | 6 - 7 | 749 | 65 |
XM_006233783.2 | XP_006233845.1 | 7 - 8 | 1915 | 65 |