Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fyttd1
Entrez Gene ID
360726
Gene Name
forty-two-three domain containing 1
Gene Aliases
Ac1176, RGD1306899
Chromosome Location
Chr.11: 71113931 - 71136721 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768736.1 XP_008766958.1 1 - 2 125 82
GenBank mRNA AY724512.1 - 1 - 2 328 82

View Full Product Details