The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207596.1 | NP_997479.1 | 2 - 3 | 175 | 105 |
GenBank mRNA | BC058156.1 | - | 2 - 3 | 175 | 105 |
FQ227408.1 | - | 2 - 3 | 176 | 105 |