Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Samd3
Entrez Gene ID
685940
Gene Name
sterile alpha motif domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 20111548 - 20155960 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172116.1 NP_001165587.1 3 - 4 375 147

View Full Product Details