Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd23
Entrez Gene ID
316330
Gene Name
ankyrin repeat domain 23
Gene Aliases
-
Chromosome Location
Chr.9: 43110980 - 43117585 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108211.1 NP_001101681.1 6 - 7 816 112
XM_006244814.2 XP_006244876.1 5 - 6 579 112

View Full Product Details