Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst10
Entrez Gene ID
140568
Gene Name
carbohydrate sulfotransferase 10
Gene Aliases
Hnk-1st
Chromosome Location
Chr.9: 45528928 - 45559184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080397.1 NP_536322.1 2 - 3 359 77
XM_006244730.2 XP_006244792.1 3 - 4 508 77
XM_006244732.2 XP_006244794.1 2 - 3 166 77
XM_008766985.1 XP_008765207.1 2 - 3 290 77
XM_008766987.1 XP_008765209.1 2 - 3 260 77
GenBank mRNA AF022729.1 - 2 - 3 359 77

View Full Product Details