Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Khdrbs2
Entrez Gene ID
170843
Gene Name
KH domain containing, RNA binding, signal transduction associated 2
Gene Aliases
Slm1
Chromosome Location
Chr.9: 39501431 - 40008702 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133318.1 NP_579852.1 2 - 3 373 135
XM_008766952.1 XP_008765174.1 2 - 3 393 135
GenBank mRNA AF305618.1 - 2 - 3 373 135

View Full Product Details