Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txndc15
Entrez Gene ID
307180
Gene Name
thioredoxin domain containing 15
Gene Aliases
RGD1304696
Chromosome Location
Chr.17: 9429171 - 9441628 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024998.1 NP_001020169.1 4 - 5 908 96
XM_008771497.1 XP_008769719.1 4 - 5 1016 96
GenBank mRNA BC091340.1 - 4 - 5 908 96

View Full Product Details