Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrs1
Entrez Gene ID
297784
Gene Name
RRS1 ribosome biogenesis regulator homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.5: 9430710 - 9432343 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079699.1 NP_001073167.1 1 - 1 1559 136
GenBank mRNA BC128746.1 - 1 - 1 1559 136

View Full Product Details