Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl5
Entrez Gene ID
305424
Gene Name
F-box and leucine-rich repeat protein 5
Gene Aliases
-
Chromosome Location
Chr.14: 71852649 - 71891143 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107222.1 NP_001100692.1 3 - 4 374 143
XM_006251076.2 XP_006251138.1 3 - 4 455 143

View Full Product Details