Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd7
Entrez Gene ID
293996
Gene Name
PDZ domain containing 7
Gene Aliases
Pdzk7
Chromosome Location
Chr.1: 264776398 - 264796206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106362.2 NP_001099832.2 6 - 7 1063 68
XM_006231449.2 XP_006231511.1 7 - 8 1669 68
XM_008760406.1 XP_008758628.1 7 - 8 1669 68
XM_008760407.1 XP_008758629.1 7 - 8 1669 68

View Full Product Details