The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107598.1 | NP_001101068.1 | 1 - 2 | 152 | 161 |
XM_008760428.1 | XP_008758650.1 | 1 - 2 | 185 | 161 |