Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmie
Entrez Gene ID
501061
Gene Name
transmembrane inner ear
Gene Aliases
RGD1562523
Chromosome Location
Chr.8: 119142113 - 119157071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109299.1 NP_001102769.1 1 - 2 804 70

View Full Product Details