The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013110.1 | NP_001013128.1 | 16 - 17 | 2096 | 99 |
GenBank mRNA | AF476964.1 | - | 17 - 18 | 2077 | 99 |
AY327504.1 | - | 16 - 17 | 2065 | 99 | |
BC087021.1 | - | 16 - 17 | 2096 | 99 | |
D38380.1 | - | 16 - 17 | 2095 | 99 | |
FQ209524.1 | - | 9 - 10 | 1156 | 99 | |
FQ209552.1 | - | 7 - 8 | 875 | 99 | |
FQ209673.1 | - | 10 - 11 | 1263 | 99 | |
FQ209674.1 | - | 10 - 11 | 1321 | 99 | |
FQ209794.1 | - | 8 - 9 | 948 | 99 | |
FQ210028.1 | - | 5 - 6 | 681 | 99 | |
FQ210115.1 | - | 4 - 5 | 576 | 99 | |
FQ210327.1 | - | 4 - 5 | 552 | 99 | |
FQ210637.1 | - | 4 - 5 | 494 | 99 | |
FQ210903.1 | - | 1 - 2 | 60 | 99 | |
FQ210963.1 | - | 5 - 6 | 620 | 99 | |
FQ211385.1 | - | 7 - 8 | 872 | 99 | |
FQ211460.1 | - | 2 - 3 | 378 | 99 | |
FQ214362.1 | - | 9 - 10 | 1173 | 99 | |
FQ219264.1 | - | 10 - 11 | 1199 | 99 | |
FQ219315.1 | - | 5 - 6 | 737 | 99 | |
FQ219349.1 | - | 8 - 9 | 929 | 99 | |
FQ219354.1 | - | 2 - 3 | 687 | 99 | |
FQ219664.1 | - | 5 - 6 | 590 | 99 | |
FQ219724.1 | - | 10 - 11 | 1228 | 99 | |
X77158.1 | - | 16 - 17 | 2073 | 99 |