Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss42
Entrez Gene ID
301027
Gene Name
protease, serine, 42
Gene Aliases
RGD1562548, Tessp2
Chromosome Location
Chr.8: 119064961 - 119070160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106863.2 NP_001100333.2 4 - 5 804 59

View Full Product Details