Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stac
Entrez Gene ID
363152
Gene Name
SH3 and cysteine rich domain
Gene Aliases
-
Chromosome Location
Chr.8: 119820664 - 119941829 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_343491.7 XP_343492.4 2 - 3 630 62

View Full Product Details