Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Col6a6
Entrez Gene ID
315979
Gene Name
collagen, type VI, alpha 6
Gene Aliases
RGD1309172
Chromosome Location
Chr.8: 114353563 - 114499373 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766605.1 XP_008764827.1 3 - 4 1838 70

View Full Product Details