Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa5
Entrez Gene ID
301013
Gene Name
shisa family member 5
Gene Aliases
-
Chromosome Location
Chr.8: 117781014 - 117795923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243761.1 XP_006243823.1 2 - 3 336 124
GenBank mRNA FQ231597.1 - 2 - 3 323 124

View Full Product Details