Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc2
Entrez Gene ID
301033
Gene Name
leucine rich repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.8: 119228533 - 119259649 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012001.1 NP_001012001.1 2 - 3 190 65
GenBank mRNA BC085884.1 - 2 - 3 190 65

View Full Product Details