Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Panx1
Entrez Gene ID
315435
Gene Name
Pannexin 1
Gene Aliases
px1
Chromosome Location
Chr.8: 13567185 - 13606040 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270548.1 NP_001257477.1 3 - 4 1428 83
NM_199397.2 NP_955429.1 4 - 5 1638 83
GenBank mRNA AJ557015.1 - 4 - 5 1638 83
GQ499839.1 - 3 - 4 1092 83

View Full Product Details