Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chordc1
Entrez Gene ID
315447
Gene Name
cysteine and histidine-rich domain (CHORD)-containing 1
Gene Aliases
CHP-1
Chromosome Location
Chr.8: 17421552 - 17445513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108128.1 NP_001101598.1 8 - 9 814 79

View Full Product Details