Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acp5
Entrez Gene ID
25732
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
TTRRAP, Trap
Chromosome Location
Chr.8: 23142733 - 23149092 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242691.2 XP_006242753.1 1 - 2 619 66

View Full Product Details