Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctla2a
Entrez Gene ID
498690
Gene Name
cytotoxic T lymphocyte-associated protein 2 alpha
Gene Aliases
RGD1565540
Chromosome Location
Chr.17: 4169695 - 4194700 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109115.1 NP_001102585.1 4 - 5 469 68
GenBank mRNA FQ220143.1 - 3 - 4 710 68

View Full Product Details