The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015023.1 | NP_001015023.1 | 3 - 4 | 691 | 69 |
XM_006253632.2 | XP_006253694.1 | 3 - 4 | 582 | 69 | |
GenBank mRNA | BC091184.1 | - | 3 - 4 | 691 | 69 |