Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inpp5j
Entrez Gene ID
171088
Gene Name
inositol polyphosphate-5-phosphatase J
Gene Aliases
Inpp, Pib5pa, Pipp
Chromosome Location
Chr.14: 83730861 - 83742011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133562.1 NP_598246.1 1 - 2 153 129
XM_008770248.1 XP_008768470.1 1 - 2 195 129
GenBank mRNA AB032551.1 - 1 - 2 153 129

View Full Product Details