Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Morc2
Entrez Gene ID
289736
Gene Name
MORC family CW-type zinc finger 2
Gene Aliases
Morc2a, Zcwcc1
Chromosome Location
Chr.14: 83889140 - 83930263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106016.1 NP_001099486.1 15 - 16 1611 73
XM_006251198.2 XP_006251260.1 17 - 18 1754 73

View Full Product Details