Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
289443
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
RGD1306585
Chromosome Location
Chr.14: 5283655 - 5378698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037179.1 NP_001032256.1 1 - 2 199 131
GenBank mRNA BC100076.1 - 1 - 2 199 131

View Full Product Details