The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037179.1 | NP_001032256.1 | 2 - 3 | 271 | 125 |
GenBank mRNA | BC100076.1 | - | 2 - 3 | 271 | 125 |