Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oat
Entrez Gene ID
64313
Gene Name
ornithine aminotransferase
Gene Aliases
rOAT
Chromosome Location
Chr.1: 204562289 - 204582070 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022521.3 NP_071966.1 7 - 8 1005 128
GenBank mRNA BC061551.1 - 7 - 8 1008 128
FQ210816.1 - 1 - 2 106 128
M11842.1 - 6 - 7 948 128

View Full Product Details