The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022521.3 | NP_071966.1 | 9 - 10 | 1267 | 80 |
GenBank mRNA | BC061551.1 | - | 9 - 10 | 1270 | 80 |
FQ210816.1 | - | 3 - 4 | 368 | 80 | |
M11842.1 | - | 8 - 9 | 1210 | 80 |