The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011940.1 | NP_001011940.1 | 7 - 8 | 1021 | 116 |
XM_006231029.2 | XP_006231091.1 | 7 - 8 | 1061 | 116 | |
GenBank mRNA | BC079048.1 | - | 7 - 8 | 1021 | 116 |