Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnni2
Entrez Gene ID
29389
Gene Name
troponin I type 2 (skeletal, fast)
Gene Aliases
-
Chromosome Location
Chr.1: 215609110 - 215611652 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017185.1 NP_058881.1 5 - 6 308 119
GenBank mRNA FQ216337.1 - 5 - 6 350 119
FQ217026.1 - 6 - 7 350 119
FQ217309.1 - 5 - 6 341 119
FQ217454.1 - 6 - 7 358 119
FQ217985.1 - 6 - 7 350 119
FQ223581.1 - 6 - 7 350 119
FQ223772.1 - 6 - 7 356 119
FQ224443.1 - 6 - 7 351 119
FQ224464.1 - 6 - 7 350 119
FQ224489.1 - 6 - 7 350 119
M73701.1 - 5 - 6 308 119

View Full Product Details