Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a10
Entrez Gene ID
293739
Gene Name
membrane-spanning 4-domains, subfamily A, member 10
Gene Aliases
-
Chromosome Location
Chr.1: 227016795 - 227030220 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106336.1 NP_001099806.1 4 - 5 431 81
XM_006231032.2 XP_006231094.1 4 - 5 708 81

View Full Product Details