The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199095.1 | NP_954526.1 | 2 - 3 | 2447 | 102 |
XM_006236459.1 | XP_006236521.1 | 3 - 4 | 2189 | 102 | |
GenBank mRNA | D00921.1 | - | 2 - 3 | 1679 | 102 |