Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubqln4
Entrez Gene ID
310633
Gene Name
ubiquilin 4
Gene Aliases
RGD1308273
Chromosome Location
Chr.2: 187915701 - 187931035 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107688.1 NP_001101158.1 7 - 8 1345 68
XM_006232642.1 XP_006232704.1 6 - 7 1139 68

View Full Product Details