Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fscn3
Entrez Gene ID
296947
Gene Name
fascin actin-bundling protein 3, testicular
Gene Aliases
-
Chromosome Location
Chr.4: 55720020 - 55729442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004232.1 NP_001004232.1 5 - 6 1394 97
GenBank mRNA BC079410.1 - 5 - 6 1394 97

View Full Product Details