Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eef2
Entrez Gene ID
29565
Gene Name
eukaryotic translation elongation factor 2
Gene Aliases
Ef-2
Chromosome Location
Chr.7: 11401501 - 11406771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017245.2 NP_058941.1 8 - 9 1671 63
GenBank mRNA BC066661.1 - 8 - 9 1671 63
K03502.1 - 1 - 2 65 63
Y07504.1 - 8 - 9 1657 63

View Full Product Details